Assay Details
Gene Symbol: LAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119651.4 | NP_567971.1 | 1 - 2 | 335 | 84 | |
GenBank mRNA | AY085918.1 | - | 1 - 2 | 226 | 84 | |
DQ062353.1 | - | 1 - 2 | 190 | 84 | ||
DQ062354.1 | - | 1 - 2 | 190 | 84 | ||
DQ062355.1 | - | 1 - 2 | 190 | 84 | ||
DQ062356.1 | - | 1 - 2 | 190 | 84 | ||
DQ062357.1 | - | 1 - 2 | 190 | 84 | ||
DQ062359.1 | - | 1 - 2 | 190 | 84 | ||
DQ062360.1 | - | 1 - 2 | 190 | 84 | ||
DQ062362.1 | - | 1 - 2 | 190 | 84 | ||
DQ062365.1 | - | 1 - 2 | 190 | 84 | ||
DQ062366.1 | - | 1 - 2 | 190 | 84 | ||
DQ062369.1 | - | 1 - 2 | 190 | 84 | ||
DQ062370.1 | - | 1 - 2 | 190 | 84 | ||
DQ062371.1 | - | 1 - 2 | 190 | 84 | ||
DQ062372.1 | - | 1 - 2 | 190 | 84 | ||
DQ062373.1 | - | 1 - 2 | 190 | 84 | ||
DQ062374.1 | - | 1 - 2 | 190 | 84 | ||
DQ062375.1 | - | 1 - 2 | 190 | 84 | ||
DQ062377.1 | - | 1 - 2 | 190 | 84 | ||
DQ062378.1 | - | 1 - 2 | 190 | 84 | ||
DQ062379.1 | - | 1 - 2 | 190 | 84 | ||
DQ062381.1 | - | 1 - 2 | 190 | 84 | ||
DQ062382.1 | - | 1 - 2 | 190 | 84 | ||
DQ062383.1 | - | 1 - 2 | 190 | 84 | ||
DQ062384.1 | - | 1 - 2 | 190 | 84 | ||
DQ446894.1 | - | 1 - 2 | 190 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
