Assay Details
| Gene Symbol: XDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119655.4 | NP_195215.2 | 9 - 10 | 3343 | 81 | |
| GenBank mRNA | AY171562.1 | - | 9 - 10 | 3140 | 81 | |
More Information
Set Membership: |
|

XDH1
Genomic Map