Assay Details
| Gene Symbol: APX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119666.4 | NP_195226.1 | 1 - 2 | 296 | 70 | |
| GenBank mRNA | AY065143.1 | - | 1 - 2 | 204 | 70 | |
| AY081646.1 | - | 1 - 2 | 116 | 70 | ||
| AY086162.1 | - | 1 - 2 | 230 | 70 | ||
| U69138.1 | - | 1 - 2 | 242 | 70 | ||
| X98003.1 | - | 186 | 70 | |||
| X98276.1 | - | 122 | 70 | |||
More Information
Set Membership: |
|

APX3
Genomic Map