Assay Details
| Gene Symbol: APX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119666.4 | NP_195226.1 | 6 - 7 | 667 | 101 | |
| GenBank mRNA | AY065143.1 | - | 6 - 7 | 575 | 101 | |
| AY081646.1 | - | 6 - 7 | 487 | 101 | ||
| AY086162.1 | - | 6 - 7 | 601 | 101 | ||
| U69138.1 | - | 6 - 7 | 613 | 101 | ||
| X98003.1 | - | 557 | 101 | |||
| X98276.1 | - | 493 | 101 | |||
More Information
Set Membership: |
|

APX3
Genomic Map