Assay Details
| Gene Symbol: BGAL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119667.3 | NP_567973.1 | 4 - 5 | 702 | 132 | |
| GenBank mRNA | AJ270307.1 | - | 4 - 5 | 591 | 132 | |
More Information
Set Membership: |
|

BGAL11
Genomic Map