Assay Details
| Gene Symbol: FAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198471.1 | NP_001185400.1 | 4 - 5 | 971 | 138 | |
| NM_106154.4 | NP_565097.1 | 3 - 4 | 1121 | 138 | ||
| NM_179557.4 | NP_849888.1 | 3 - 4 | 1121 | 138 | ||
| GenBank mRNA | AF318307.1 | - | 3 - 4 | 806 | 138 | |
| AF419598.1 | - | 3 - 4 | 885 | 138 | ||
| AY054196.1 | - | 3 - 4 | 966 | 138 | ||
| AY097344.1 | - | 3 - 4 | 719 | 138 | ||
More Information
Set Membership: |
|

FAB1
Genomic Map