Assay Details
Gene Symbol: GGH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106513.5 | NP_177987.1 | 3 - 4 | 514 | 116 | |
NM_179569.4 | NP_849900.1 | 2 - 3 | 485 | 116 | ||
NM_202443.3 | NP_974172.1 | 4 - 5 | 545 | 116 | ||
GenBank mRNA | AK317198.1 | - | 2 - 3 | 471 | 116 | |
BT024737.1 | - | 2 - 3 | 407 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
