Assay Details
| Gene Symbol: GGH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106513.5 | NP_177987.1 | 4 - 5 | 612 | 106 | |
| NM_179569.4 | NP_849900.1 | 3 - 4 | 583 | 106 | ||
| NM_202443.3 | NP_974172.1 | 5 - 6 | 643 | 106 | ||
| GenBank mRNA | AK317198.1 | - | 3 - 4 | 569 | 106 | |
| BT024737.1 | - | 3 - 4 | 505 | 106 | ||
More Information
Set Membership: |
|

GGH1
Genomic Map