Assay Details
| Gene Symbol: PGK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106603.3 | NP_178073.1 | 1 - 2 | 380 | 87 | |
| NM_179576.3 | NP_849907.1 | 2 - 3 | 397 | 87 | ||
| GenBank mRNA | AF247560.1 | - | 1 - 2 | 225 | 87 | |
| AF348582.1 | - | 1 - 2 | 191 | 87 | ||
| AY062863.1 | - | 2 - 3 | 298 | 87 | ||
| BT008486.1 | - | 1 - 2 | 191 | 87 | ||
More Information
Set Membership: |
|

PGK
Genomic Map