Assay Details
| Gene Symbol: TPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334989.1 | NP_001319429.1 | 16 - 17 | 2357 | 73 | |
| NM_106697.5 | NP_178164.3 | 16 - 17 | 2348 | 73 | ||
| NM_179582.2 | NP_849913.2 | 16 - 17 | 2302 | 73 | ||
More Information
Set Membership: |
|

TPR1
Genomic Map