Assay Details
Gene Symbol: TPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334989.1 | NP_001319429.1 | 21 - 22 | 3081 | 84 | |
NM_106697.5 | NP_178164.3 | 21 - 22 | 3072 | 84 | ||
NM_179582.2 | NP_849913.2 | 21 - 22 | 3026 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
