Assay Details
Gene Symbol: CALS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179622.4 | NP_849953.2 | 14 - 15 | 1521 | 87 | |
GenBank mRNA | AY337762.1 | - | 14 - 15 | 1504 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
