Assay Details
Gene Symbol: PNP-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179648.2 | NP_849979.1 | 1 - 2 | 210 | 78 | |
GenBank mRNA | AY065269.1 | - | 1 - 2 | 130 | 78 | |
AY142603.1 | - | 1 - 2 | 118 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
