Assay Details
Gene Symbol: PNP-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179648.2 | NP_849979.1 | 2 - 3 | 481 | 93 | |
GenBank mRNA | AY065269.1 | - | 2 - 3 | 401 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
