Assay Details
| Gene Symbol: HPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179653.4 | NP_849984.1 | 10 - 11 | 1229 | 82 | |
| GenBank mRNA | AF324344.1 | - | 10 - 11 | 1051 | 82 | |
| AY063893.1 | - | 10 - 11 | 1081 | 82 | ||
| AY089963.1 | - | 10 - 11 | 937 | 82 | ||
| AY113993.1 | - | 10 - 11 | 937 | 82 | ||
| GU198365.1 | - | 10 - 11 | 1015 | 82 | ||
More Information
Set Membership: |
|

HPT1
Genomic Map