Assay Details
| Gene Symbol: HPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335615.1 | NP_001325303.1 | 2 - 3 | 257 | 74 | |
| NM_179653.4 | NP_849984.1 | 2 - 3 | 378 | 74 | ||
| GenBank mRNA | AF324344.1 | - | 2 - 3 | 200 | 74 | |
| AY063893.1 | - | 2 - 3 | 230 | 74 | ||
| AY089963.1 | - | 2 - 3 | 86 | 74 | ||
| AY113993.1 | - | 2 - 3 | 86 | 74 | ||
| GU198365.1 | - | 2 - 3 | 164 | 74 | ||
More Information
Set Membership: |
|

HPT1
Genomic Map