Assay Details
| Gene Symbol: HPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335615.1 | NP_001325303.1 | 4 - 5 | 592 | 124 | |
| NM_179653.4 | NP_849984.1 | 4 - 5 | 713 | 124 | ||
| GenBank mRNA | AF324344.1 | - | 4 - 5 | 535 | 124 | |
| AY063893.1 | - | 4 - 5 | 565 | 124 | ||
| AY089963.1 | - | 4 - 5 | 421 | 124 | ||
| AY113993.1 | - | 4 - 5 | 421 | 124 | ||
| GU198365.1 | - | 4 - 5 | 499 | 124 | ||
More Information
Set Membership: |
|

HPT1
Genomic Map