Assay Details
| Gene Symbol: HPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335615.1 | NP_001325303.1 | 7 - 8 | 846 | 90 | |
| NM_179653.4 | NP_849984.1 | 7 - 8 | 967 | 90 | ||
| GenBank mRNA | AF324344.1 | - | 7 - 8 | 789 | 90 | |
| AY063893.1 | - | 7 - 8 | 819 | 90 | ||
| AY089963.1 | - | 7 - 8 | 675 | 90 | ||
| AY113993.1 | - | 7 - 8 | 675 | 90 | ||
| GU198365.1 | - | 7 - 8 | 753 | 90 | ||
More Information
Set Membership: |
|

HPT1
Genomic Map