Assay Details
| Gene Symbol: DWA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179658.4 | NP_849989.1 | 7 - 8 | 860 | 89 | |
| GenBank mRNA | AY099718.1 | - | 7 - 8 | 713 | 89 | |
| AY128879.1 | - | 7 - 8 | 689 | 89 | ||
More Information
Set Membership: |
|

DWA1
Genomic Map