Assay Details
| Gene Symbol: SR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124889.2 | NP_001118361.1 | 3 - 4 | 467 | 120 | |
| NM_001335795.1 | NP_001325089.1 | 2 - 3 | 993 | 120 | ||
| NM_179692.3 | NP_850023.1 | 2 - 3 | 539 | 120 | ||
| GenBank mRNA | AF506697.1 | - | 2 - 3 | 163 | 120 | |
| AY510025.1 | - | 2 - 3 | 163 | 120 | ||
| BT002459.1 | - | 2 - 3 | 346 | 120 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001335795.1 transcripts.
Set Membership: |
|

SR1
Genomic Map