Assay Details
| Gene Symbol: SR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124889.2 | NP_001118361.1 | 7 - 8 | 923 | 86 | |
| NM_001335795.1 | NP_001325089.1 | 6 - 7 | 1449 | 86 | ||
| NM_179692.3 | NP_850023.1 | 6 - 7 | 995 | 86 | ||
| GenBank mRNA | AF506697.1 | - | 6 - 7 | 619 | 86 | |
| AY510025.1 | - | 6 - 7 | 619 | 86 | ||
| BT002459.1 | - | 6 - 7 | 802 | 86 | ||
More Information
Set Membership: |
|

SR1
Genomic Map