Assay Details
| Gene Symbol: SR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124889.2 | NP_001118361.1 | 10 - 11 | 1384 | 117 | |
| NM_001335795.1 | NP_001325089.1 | 9 - 10 | 1910 | 117 | ||
| NM_179692.3 | NP_850023.1 | 9 - 10 | 1456 | 117 | ||
| GenBank mRNA | AF506697.1 | - | 9 - 10 | 1080 | 117 | |
| AY510025.1 | - | 9 - 10 | 1080 | 117 | ||
| BT002459.1 | - | 9 - 10 | 1263 | 117 | ||
More Information
Set Membership: |
|

SR1
Genomic Map