Assay Details
| Gene Symbol: ARF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084486.1 | NP_001077955.1 | 5 - 6 | 431 | 64 | |
| NM_179726.3 | NP_850057.1 | 5 - 6 | 469 | 64 | ||
| GenBank mRNA | AK117735.1 | - | 5 - 6 | 412 | 64 | |
| AY063840.1 | - | 5 - 6 | 431 | 64 | ||
| AY150410.1 | - | 5 - 6 | 335 | 64 | ||
| X77385.1 | - | 363 | 64 | |||
More Information
Set Membership: |
|

ARF3
Genomic Map