Assay Details
| Gene Symbol: FRS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036351.2 | NP_001031428.1 | 4 - 5 | 2208 | 65 | |
| NM_128269.4 | NP_565636.1 | 3 - 4 | 2505 | 65 | ||
| NM_179767.2 | NP_850098.1 | 2 - 3 | 2652 | 65 | ||
| GenBank mRNA | AK317350.1 | - | 4 - 5 | 2151 | 65 | |
| AY057491.1 | - | 3 - 4 | 2443 | 65 | ||
| BT002690.1 | - | 2 - 3 | 2240 | 65 | ||
More Information
Set Membership: |
|

FRS3
Genomic Map