Assay Details
Gene Symbol: NLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336116.1 | NP_001324215.1 | 1 - 2 | 472 | 90 | |
NM_128305.2 | NP_565650.1 | 2 - 3 | 255 | 90 | ||
NM_179770.1 | NP_850101.1 | 2 - 3 | 244 | 90 | ||
GenBank mRNA | AK317300.1 | - | 2 - 3 | 175 | 90 | |
AY072113.1 | - | 2 - 3 | 244 | 90 | ||
AY086056.1 | - | 2 - 3 | 255 | 90 | ||
AY122963.1 | - | 2 - 3 | 223 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001336116.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
