Assay Details
| Gene Symbol: NLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336116.1 | NP_001324215.1 | 2 - 3 | 571 | 68 | |
| NM_128305.2 | NP_565650.1 | 3 - 4 | 354 | 68 | ||
| NM_179770.1 | NP_850101.1 | 3 - 4 | 343 | 68 | ||
| GenBank mRNA | AK317300.1 | - | 3 - 4 | 274 | 68 | |
| AY072113.1 | - | 3 - 4 | 343 | 68 | ||
| AY086056.1 | - | 3 - 4 | 354 | 68 | ||
| AY122963.1 | - | 3 - 4 | 322 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001336116.1 transcripts.
Set Membership: |
|

NLP1
Genomic Map