Assay Details
| Gene Symbol: NLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336116.1 | NP_001324215.1 | 3 - 4 | 714 | 104 | |
| NM_128305.2 | NP_565650.1 | 4 - 5 | 497 | 104 | ||
| NM_179770.1 | NP_850101.1 | 4 - 5 | 486 | 104 | ||
| GenBank mRNA | AK317300.1 | - | 4 - 5 | 417 | 104 | |
| AY072113.1 | - | 4 - 5 | 486 | 104 | ||
| AY086056.1 | - | 4 - 5 | 497 | 104 | ||
| AY122963.1 | - | 4 - 5 | 465 | 104 | ||
More Information
Set Membership: |
|

NLP1
Genomic Map