Assay Details
Gene Symbol: FRA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179782.4 | NP_850113.2 | 2 - 3 | 994 | 128 | |
GenBank mRNA | DQ182567.1 | - | 2 - 3 | 673 | 128 | |
KJ138894.1 | - | 2 - 3 | 632 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
