Assay Details
Gene Symbol: BGAL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202695.2 | NP_001189624.1 | 20 - 21 | 3021 | 68 | |
NM_001336165.1 | NP_001324192.1 | 18 - 19 | 3272 | 68 | ||
NM_001336166.1 | NP_001324193.1 | 19 - 20 | 3125 | 68 | ||
NM_179790.3 | NP_850121.1 | 19 - 20 | 3168 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
