Assay Details
Gene Symbol: BGAL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202695.2 | NP_001189624.1 | 8 - 9 | 1273 | 107 | |
NM_001336165.1 | NP_001324192.1 | 6 - 7 | 1524 | 107 | ||
NM_001336166.1 | NP_001324193.1 | 7 - 8 | 1377 | 107 | ||
NM_001336167.1 | NP_001324194.1 | 6 - 7 | 1212 | 107 | ||
NM_179790.3 | NP_850121.1 | 7 - 8 | 1420 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
