Assay Details
| Gene Symbol: emb1997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179796.2 | NP_850127.1 | 8 - 9 | 1294 | 146 | |
| GenBank mRNA | AX765866.1 | - | 7 - 8 | 1069 | 146 | |
| AY096797.1 | - | 8 - 9 | 1201 | 146 | ||
| BT002702.1 | - | 7 - 8 | 1069 | 146 | ||
More Information
Set Membership: |
|

emb1997
Genomic Map