Assay Details
| Gene Symbol: ETC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179814.4 | NP_850145.1 | 1 - 2 | 334 | 133 | |
| GenBank mRNA | AY234411.1 | - | 1 - 2 | 334 | 133 | |
| AY519520.1 | - | 1 - 2 | 87 | 133 | ||
| AY649255.1 | - | 1 - 2 | 87 | 133 | ||
More Information
Set Membership: |
|

ETC2
Genomic Map