Assay Details
Gene Symbol: HSI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179815.3 | NP_850146.1 | 6 - 7 | 1681 | 84 | |
GenBank mRNA | AB206553.1 | - | 6 - 7 | 1436 | 84 | |
AY062492.1 | - | 6 - 7 | 1444 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
