Assay Details
| Gene Symbol: LGT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179819.2 | NP_850150.1 | 7 - 8 | 1351 | 95 | |
| GenBank mRNA | AY081313.1 | - | 7 - 8 | 1344 | 95 | |
| BT002563.1 | - | 7 - 8 | 1039 | 95 | ||
| KJ138886.1 | - | 7 - 8 | 1039 | 95 | ||
More Information
Set Membership: |
|

LGT5
Genomic Map