Assay Details
Gene Symbol: MT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112401.2 | NP_566509.1 | 1 - 2 | 263 | 59 | |
GenBank mRNA | AK229545.1 | - | 1 - 2 | 133 | 59 | |
AY074340.1 | - | 1 - 2 | 130 | 59 | ||
AY087213.1 | - | 1 - 2 | 127 | 59 | ||
AY091386.1 | - | 1 - 2 | 51 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
