Assay Details
| Gene Symbol: UBC25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112402.3 | NP_188154.1 | 4 - 5 | 1259 | 124 | |
| GenBank mRNA | AF361806.1 | - | 4 - 5 | 1161 | 124 | |
| AY059155.1 | - | 4 - 5 | 1042 | 124 | ||
More Information
Set Membership: |
|

UBC25
Genomic Map