Assay Details
Gene Symbol: NAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112419.4 | NP_188170.1 | 2 - 3 | 760 | 86 | |
GenBank mRNA | AB049071.1 | - | 2 - 3 | 496 | 86 | |
BT004079.1 | - | 2 - 3 | 681 | 86 | ||
BT005044.1 | - | 2 - 3 | 496 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
