Assay Details
Gene Symbol: CDC27a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338228.1 | NP_001319567.1 | 12 - 13 | 1732 | 82 | |
NM_001338229.1 | NP_001325497.1 | 12 - 13 | 1627 | 82 | ||
NM_001338230.1 | NP_001325498.1 | 12 - 13 | 1600 | 82 | ||
NM_112503.5 | NP_188253.3 | 12 - 13 | 1699 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
