Assay Details
| Gene Symbol: AT3G16600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112533.1 | NP_188282.1 | 7 - 8 | 1523 | 82 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G16600| Gene Symbol: AT3G16600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112533.1 | NP_188282.1 | 7 - 8 | 1523 | 82 | |
Set Membership: | 
		   
					
						
							 |