Assay Details
| Gene Symbol: PLDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338256.1 | NP_001326710.1 | 18 - 19 | 3558 | 77 | |
| NM_001338259.1 | NP_001326707.1 | 17 - 18 | 3725 | 77 | ||
| NM_112553.4 | NP_188302.2 | 19 - 20 | 3479 | 77 | ||
| GenBank mRNA | AF411833.1 | - | 19 - 20 | 3286 | 77 | |
More Information
Set Membership: |
|

PLDP1
Genomic Map