Assay Details
| Gene Symbol: PLDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338255.1 | NP_001326711.1 | 8 - 9 | 1728 | 129 | |
| NM_001338256.1 | NP_001326710.1 | 8 - 9 | 1728 | 129 | ||
| NM_001338257.1 | NP_001326709.1 | 8 - 9 | 1682 | 129 | ||
| NM_001338258.1 | NP_001326708.1 | 8 - 9 | 1667 | 129 | ||
| NM_001338259.1 | NP_001326707.1 | 8 - 9 | 1667 | 129 | ||
| NM_112553.4 | NP_188302.2 | 8 - 9 | 1728 | 129 | ||
| GenBank mRNA | AF411833.1 | - | 8 - 9 | 1535 | 129 | |
More Information
Set Membership: |
|

PLDP1
Genomic Map