Assay Details
| Gene Symbol: UBC32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112576.3 | NP_566563.1 | 3 - 4 | 700 | 70 | |
| GenBank mRNA | AF386935.1 | - | 3 - 4 | 450 | 70 | |
| BT020530.1 | - | 3 - 4 | 334 | 70 | ||
More Information
Set Membership: |
|

UBC32
Genomic Map