Assay Details
| Gene Symbol: AT3G17060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112582.2 | NP_188331.1 | 3 - 4 | 658 | 70 | |
| GenBank mRNA | AK118362.1 | - | 3 - 4 | 647 | 70 | |
| AY085221.1 | - | 3 - 4 | 658 | 70 | ||
| BT005685.1 | - | 3 - 4 | 618 | 70 | ||
More Information
Set Membership: |
|

AT3G17060
Genomic Map