Assay Details
| Gene Symbol: POK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338286.1 | NP_001326050.1 | 31 - 32 | 6084 | 106 | |
| NM_001338287.1 | NP_001326049.1 | 31 - 32 | 6043 | 106 | ||
| NM_112614.4 | NP_188362.2 | 31 - 32 | 6060 | 106 | ||
| GenBank mRNA | DQ399529.1 | - | 31 - 32 | 5832 | 106 | |
More Information
Set Membership: |
|

POK1
Genomic Map