Assay Details
Gene Symbol: XPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112681.3 | NP_188427.2 | 9 - 10 | 1455 | 114 | |
GenBank mRNA | AF197940.1 | - | 9 - 10 | 1441 | 114 | |
AF367299.1 | - | 9 - 10 | 1380 | 114 | ||
AY058175.1 | - | 9 - 10 | 1455 | 114 | ||
AY091683.1 | - | 9 - 10 | 1240 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
