Assay Details
| Gene Symbol: BGLU43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338322.1 | NP_001325994.1 | 4 - 5 | 447 | 114 | |
| NM_001338323.1 | NP_001325998.1 | 4 - 5 | 420 | 114 | ||
| NM_112689.2 | NP_188435.2 | 4 - 5 | 420 | 114 | ||
More Information
Set Membership: |
|

BGLU43
Genomic Map