Assay Details
| Gene Symbol: BGLU44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112690.4 | NP_188436.1 | 4 - 5 | 569 | 115 | |
| GenBank mRNA | AK316840.1 | - | 4 - 5 | 472 | 115 | |
| AK316900.1 | - | 4 - 5 | 473 | 115 | ||
| AY084864.1 | - | 4 - 5 | 471 | 115 | ||
More Information
Set Membership: |
|

BGLU44
Genomic Map