Assay Details
| Gene Symbol: BGLU44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112690.4 | NP_188436.1 | 8 - 9 | 1253 | 98 | |
| GenBank mRNA | AK316840.1 | - | 8 - 9 | 1156 | 98 | |
| AK316900.1 | - | 8 - 9 | 1157 | 98 | ||
| AY084864.1 | - | 8 - 9 | 1155 | 98 | ||
More Information
Set Membership: |
|

BGLU44
Genomic Map