Assay Details
| Gene Symbol: ATSYTF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112722.3 | NP_566607.1 | 10 - 11 | 2456 | 73 | |
| GenBank mRNA | AY056799.1 | - | 10 - 11 | 1925 | 73 | |
| AY133527.1 | - | 10 - 11 | 1538 | 73 | ||
More Information
Set Membership: |
|

ATSYTF
Genomic Map