Assay Details
| Gene Symbol: ATSYTF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112722.3 | NP_566607.1 | 9 - 10 | 2236 | 78 | |
| GenBank mRNA | AY056799.1 | - | 9 - 10 | 1705 | 78 | |
| AY133527.1 | - | 9 - 10 | 1318 | 78 | ||
More Information
Set Membership: |
|

ATSYTF
Genomic Map